missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptokinase Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA185891
This item is not returnable.
View return policy
Description
Sheep anti Streptokinase antibody recognizes intact streptokinase, a metallo-enzyme produced by various strains of Streptococcus that can liquify blood clots by converting plasminogen to plasmin.
Streptokinase is a metallo-enzyme produced by various strains of Streptococcus that can liquify blood clots by converting plasminogen to plasmin.
Specifications
| Streptokinase | |
| Polyclonal | |
| Unconjugated | |
| Sheep | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Purified, native Streptococcus streptokinase. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction