missing translation for 'onlineSavingsMsg'
Learn More

Streptozocin, MedChemExpress

Product Code. 16401706
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16401706

Brand: Medchem Express HY13753

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Streptozocin is a potent DNA-methylating antibiotic. Streptozotocin causes methylation of liver and kidney and pancreatic DNA, but no methylation in brain DNA.

Streptozocin is a potent DNA-methylating antibiotic. Streptozotocin causes methylation of liver and kidney and pancreatic DNA, but no methylation in brain DNA.

Specifications

Chemical Name or Material Streptozocin
Physical Form Powder
CAS 18883-66-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C8H15N3O7
For Use With (Application) Cancer-programmed cell death
Synonym Streptozotocin U 9889
Solubility Information H2O : 113.3 mg/mL (427.19 mM; Need ultrasonic and warming) ∣DMSO : 83.33 mg/mL (314.19 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](CO)O[C@H](O)[C@H](NC(N(C)N=O)=O)[C@H]1O
Molecular Weight (g/mol) 265.22
Formula Weight 265.22
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.