Learn More
Tocris Bioscience™ Streptozocin
Description
Antibiotic and antitumor agent. Alkylates DNA and induces diabetes mellitus via reduction of nicotinamide adenine dinucleotide in pancreatic β-cells in vivo.
Specifications
Specifications
| Chemical Name or Material | Streptozocin |
| CAS | 18883-66-4 |
| Quantity | 100 mg |
| Molecular Formula | C8H15N3O7 |
| Purity | >95% |