missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Striated Muscle Myosin Heavy Chain Antibody (83B6) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50254JF525
This item is not returnable.
View return policy
Description
Striated Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Striated Muscle Myosin Heavy Chain in Chicken,Human,Mouse,Rabbit,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Striated Muscle Myosin Heavy Chain | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Mouse | |
| Ascites | |
| RUO | |
| Chicken, Human, Mouse, Rabbit, Rat | |
| Ascites |
| Immunohistochemistry, Western Blot | |
| 83B6 | |
| 50mM Sodium Borate | |
| Purified human skeletal muscle myosin | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction