missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRN4 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36537H
This item is not returnable.
View return policy
Description
STRN4 Polyclonal antibody specifically detects STRN4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| STRN4 | |
| Polyclonal | |
| Western Blot | |
| FLJ35594, striatin, calmodulin binding protein 4, striatin-4, ZIN, zinedin | |
| The immunogen recognized by the antibody maps to a region between residue 325 to 375 of human Striatin-4 using the numbering given in entry NP_037535.2. | |
| 0.1 mL | |
| Neuroscience | |
| 29888 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction