missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STT3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48657
This item is not returnable.
View return policy
Description
STT3B Polyclonal antibody specifically detects STT3B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| STT3B | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FLJ90106, homolog of yeast STT3, Oligosaccharyl transferase subunit STT3B, SIMPEC 2.4.1.119, source of immunodominant MHC associated peptides, Source of immunodominant MHC-associated peptides homolog, STT3, subunit of the oligosaccharyltransferase complex, homolog B (S.cerevisiae), STT3-Bdolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KAPDNRETLDHKPRVTNIFPKQKYLSKKTTKRKRGYIK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 201595 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction