Learn More
Cole-Parmer™ Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators
Product Code. 11416558
Click to view available options
For Use With (Equipment):
For 12 Tubes
For 20 Glaser
For 30 Tubes, 26 mm dia.
For 40 Microcentrifuge Tubes
For 63 Tubes, 12 mm dia.
For 63 Tubes, 16 mm dia.
Unit Size:
Each
Product Code. 11416558
Brand: Cole-Parmer™ 0007715
Only null left
Add to basket
Add to basket
Quickly and easily replace tube holder without tools using these Tube Holder Discs for Cole-Parmer™ TR-200 Series Tube Rotators.
  • Choose the tube holder(s) that accommodates your tube size(s).
Specifications
Holds Test Tubes, Blood Tubes
Tube Size 9 to 20 mm
Product Type Tube Holder
For Use With (Equipment) For 20 Glaser
No. of Positions 20 Tubes