missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stuart™ Polypropylene Bottle Top Dispenser

Designed for routine dispensing of common laboratory liquids such as dilute acids, buffers, biological solutions, polar solvents and strong alkaline solutions. Stuart™ Polypropylene Bottle Top Dispenser provides a low cost option for liquid dispensing without compromising accuracy or reliability.

Brand:  Cole-Parmer™ BD10

Additional Details : Weight : 0.36000kg

 View more versions of this product

Product Code. 12023893

  • € 241.00 / Each


Estimated Shipment: 26-08-2021
Add to basket

Description

Description

  • Economical dispensing of non-aggressive liquids
  • Low-cost replaceable piston/cylinder and valves
  • Easy cleaning and maintenance
  • Simple to operate
  • Automatic refill
Specifications

Specifications

1.2%
1 to 10 mL
Closing aqueous solutions, alkalis, dilute acids and polar solvents such as ethanol or acetone
BD10
Dispenser
Polypropylene
Adapters for 32mm and 40mm threads, antimicrobial protection
0.2 mL
Videos
SDS
Documents
Certificates
View accessory products