missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Polypropylene/Polyethylene Dispensing Cartridges

Dispensing cartridge for bottle top dispenser

Brand:  Cole-Parmer™ BD1010

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12033893

Quantity Price
1 € 54.58 / Pack of 3
10 € 51.67 / Pack of 3
Estimated Shipment: 06-09-2021

Add to basket


Description

Description

  • Bottle top dispenser based on a polypropylene/polyethylene dispensing cartridge ideal for closing aqueous solutions, alkalis, dilute acids and polar solvents such as ethanol or acetone
  • Replaceable dispensing cartridge and valves
  • 45 mm thread size with adapters for 32 mm and 40 mm threads
  • With BioCote™ antimicrobial protection
Specifications

Specifications

Dispensing Cartridge
Polypropylene, Polyethylene
3/Pk.
10 mL
Ideal For Closing Aqueous Solutions, Alkalis, Dilute Acids And Polar Solvents Such As Ethanol Or Acetone
45 mm
Videos
SDS
Documents
Certificates