Learn More
Cole-Parmer™ Stuart™ Pressmatic Polypropylene Dispensers
Product Code. 12040902
Click to view available options
Volume (Metric):
10 to 60 mL
2 to 10 mL
Unit Size:
Each
Product Code. 12040902
Brand: Cole-Parmer™ 1011283
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Accuracy 0.7%
Product Type Bottletop Dispenser
Material Polypropylene
Volume (Metric) 10 to 60 mL
For Use With (Equipment) Bottle
Increments 1 mL