Learn More
Cole-Parmer™ Sample Concentrator Needles for Cole-Parmer™ SC-200 Series Sample Concentrator
Click to view available options
Material:
PTFE
Stainless Steel
Size:
76 mm
127 mm
Unit Size:
Pack of 100
Description
- Maintain your sample concentrator with the following needles.
Specifications
Specifications
| Material | PTFE |
| Type | Sample Concentrator Needle |
| Quantity | 100 Pk. |
| Size | 127 mm |