Learn More
Invitrogen™ STX12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117123
Description
Positive Samples: Mouse kidney, Mouse brain, Rat kidney Immunogen sequence: MSYGPLDMYR NPGPSGPQLR DFSSIIQTCS GNIQRISQAT AQIKNLMSQL GTKQDSSKLQ ENLQQLQHST NQLAKETNEL LKELGSLPLP LSTSEQRQQR LQKERLMNDF SAALNNFQAV QRRVSEKEKE SIARARAGSR LSAEERQREE QLVSFDSHEE WNQMQSQEDE VAITEQDLEL IKERETAIRQ LEADILDVNQ IFKDLAMMIH DQGDLIDSIE ANVESSEVHV ERATEQLQRA AYYQKKSRKK MCILVLVLSV IILILGLIIW LVYKTK.
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
Specifications
| STX12 | |
| Polyclonal | |
| Unconjugated | |
| STX12 | |
| AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100226, 23673, 65033 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.5 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| G3V7P1, Q86Y82, Q9ER00 | |
| STX12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human STX12 (NP_8031731). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.