missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIGHT/TNFSF14 Antibody (MM0456-8C34), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11964PCP
This item is not returnable.
View return policy
Description
LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry.
Specifications
| LIGHT/TNFSF14 | |
| Monoclonal | |
| PerCP | |
| O43557 | |
| TNFSF14 | |
| Human recombinant LIGHT/TNFSF14 (extracellular domain) | |
| 0.1 mL | |
| Apoptosis | |
| 8740 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| MM0456-8C34 | |
| Flow Cytometry | |
| CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction