Learn More
STX1A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55164-1-AP-150UL
 View more versions of this product
Product Code. 18057795
  • € 410.00 / 150µL
Description
The STX1A antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human STX1A. This antibody recognizes human antigen. The STX1A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
STX1A
350 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
HPC 1, Neuron specific antigen HPC 1, p35 1, STX1, STX1A, Syntaxin 1A, syntaxin 1A (brain)
150 μL
6804
Store at -20°C. Stable for one year after shipment.