missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STYK1 Antibody (484713), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6668P
This item is not returnable.
View return policy
Description
STYK1 Monoclonal antibody specifically detects STYK1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| STYK1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 484713 | |
| Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| DKFZp761P1010, EC 2.7.10.2, NOKProtein PK-unique, serine/threonine/tyrosine kinase 1Novel oncogene with kinase domain, SuRTK106, tyrosine-protein kinase STYK1 | |
| E. coli-derived recombinant human STYK1, Asp72-Val320, Accession # Q6J9G0 | |
| 0.1 mL | |
| Protein Kinase | |
| 55359 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction