missing translation for 'onlineSavingsMsg'
Learn More

Suberyl Glycine-13C2,15N, TRC

Product Code. 30418875
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418875 10 mg
30423370 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418875 Supplier Toronto Research Chemicals Supplier No. TRCS68875710MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C2 C8 H17 15N O5
Molecular Weight (g/mol) 234.22
Synonym 8-[(Carboxymethyl)amino]-8-oxooctanoic Acid, -13C2,15N Suberylglycine-13C2,15N, Suberylglycine-13C2,15N (glycine-13C2,15N)
IUPAC Name 8-(hydroxycarbonyl(1^{13}C)methylamino)-8-oxooctanoic acid
SMILES O[13C](=O)[13CH2][15NH]C(=O)CCCCCCC(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H17NO5/c12-8(11-7-10(15)16)5-3-1-2-4-6-9(13)14/h1-7H2,(H,11,12)(H,13,14)(H,15,16)/i7+1,10+1,11+1
Formula Weight 234.114
Chemical Name or Material Suberyl Glycine-13C2,15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.