missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Subframe for UN110, Adjustable up to 130 mm
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN110 Oven
Unit Size:
Each
Description
- Adjustable in height
Specifications
Specifications
| Description | Adjustable |
| For Use With (Equipment) | UN110 Oven |
| Type | Subframe |
| Height (Metric) | Up to 130 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction