Learn More
Memmert™ Subframe for UN55, Adjustable up to 130 mm
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN55 Oven
Unit Size:
Each
Description
- Adjustable in height
Specifications
Specifications
| Description | Adjustable |
| For Use With (Equipment) | UN55 Oven |
| Type | Subframe |
| Height (Metric) | Up to 130 mm |