missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUCLG1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97325F
This item is not returnable.
View return policy
Description
SUCLG1 Polyclonal specifically detects SUCLG1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SUCLG1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8802 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| EC 6.2.1, EC 6.2.1.4, FLJ21114, FLJ43513, GALPHA, MTDPS9, SCS-alpha, succinate-CoA ligase, alpha subunit, succinate-CoA ligase, GDP-forming, alpha subunit, succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial, Succinyl-CoA synthetase subunit alpha, SUCLA1 | |
| Produced in rabbits immunized with E. coli-derived Human SUCLG1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction