missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SUCLG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30024
This item is not returnable.
View return policy
Description
SUCLG1 Polyclonal antibody specifically detects SUCLG1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SUCLG1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 6.2.1, EC 6.2.1.4, FLJ21114, FLJ43513, GALPHA, MTDPS9, SCS-alpha, succinate-CoA ligase, alpha subunit, succinate-CoA ligase, GDP-forming, alpha subunit, succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial, Succinyl-CoA synthetase subunit alpha, SUCLA1 | |
| Between 296 and 346 | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 8802 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction