Learn More
Sucrose Brix Standard Solution, 5%, Reagecon™
Product Code. 12673076 Shop All Reagecon Products
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. 12673076
Brand: Reagecon BS05