missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, for Electrophoresis, Fisher Chemical™

Catalog Number p-7073593
Click to view available options
Quantity:
500 g
3 kg
This item is not returnable. View return policy
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
This item is not returnable. View return policy
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Melting Point 190°C
Color White
pH 6.5 to 7.5
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 342.29g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'