Learn More
Witeg™ Suction Tube with Needle Valve Stopcock
Product Code. 15890965
Change view
Click to view available options
Closure Size:
Cone ST 29/32
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size
15890965 Cone ST 29/32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15890965 Supplier Witeg™ Supplier No. 1 684 029
With needle valve stopcock.
  • Suction tube
  • Cone ST 29/32
  • With lateral olive
Specifications
Type Suction tube
Material Glass
Closure Size Cone ST 29/32
Includes Needle valve stopcock