missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sudan I Recombinant Rabbit Monoclonal Antibody (Huam029)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544665
This item is not returnable.
View return policy
Description
Application notes: Indirect ELISA.
Sudan I (also commonly known as CI Solvent Yellow 14 and Solvent Orange R), is an organic compound, typically classified as an azo dye. Sudan I is one of the industry dyes and widely used in cosmetics, wax agent, solvent and textile. Sudan I has multiple toxicity such as carcinogenicity, mutagenicity, genotoxicity and oxidative damage. It is an intensely orange-red solid that is added to colourise waxes, oils, petrol, solvents, and polishes. Sudan I has also been adopted for colouring various foodstuffs, especially curry powder and chili powder, although the use of Sudan I in foods is now banned in many countries, because Sudan I, Sudan III, and Sudan IV have been classified as category 3 carcinogens (not classifiable as to its carcinogenicity to humans) by the International Agency for Research on Cancer. Sudan I is still used in some orange-coloured smoke formulations and as a colouring for cotton refuse used in chemistry experiments. Sudan 1 is a compound being warned of for health hazards by the EU regulation. It may cause allergic skin reactions and irritation of the skin. Exposure to the skin can happen by direct exposure to textile workers or by wearing tight-fitting textiles dyed with Sudan 1. Allergic reactions are induced when the azo dye binds to the human serum albumin (HSA), forming a dye-HSA conjugate, which immunoglobulin E binds to, which causes a release of histamine.
Specifications
| Sudan I | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| Huam029 | |
| Unconjugated | |
| CI Solvent Yellow 14; Solvent Orange R | |
| Sudan I-OVA. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction