Learn More
SUFU Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-26759-100UL
Description
The SUFU antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SUFU. This antibody recognizes human, mouse, rat antigen. The SUFU antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Spécification
| SUFU | |
| 1000 μg/mL | |
| PRO1280, SUFU, SUFUH, SUFUXL, Suppressor of fused homolog | |
| 100 μL | |
| 51684 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.