Learn More
SUFU Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-26759-100UL
 Afficher plus de versions de ce produit
Code produit. 18090606
  • € 450.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
The SUFU antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SUFU. This antibody recognizes human, mouse, rat antigen. The SUFU antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Spécification
Spécification
SUFU
1000 μg/mL
PRO1280, SUFU, SUFUH, SUFUXL, Suppressor of fused homolog
100 μL
51684
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa