missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUGT1 Antibody (6G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00010910-M03
This item is not returnable.
View return policy
Description
SUGT1 Monoclonal antibody specifically detects SUGT1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
Specifications
| SUGT1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Protein 40-6-3, putative 40-6-3 protein, Sgt1, SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae), SGT1B protein, suppressor of G2 allele of SKP1 homolog, suppressor of G2 allele of SKP1, S. cerevisiae, homolog of | |
| SUGT1 (NP_006695, 151 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VVITLMIKNVQKNDVNVEFSEKELSALVKLPSGEDYNLKLELLHPIIPEQSTFKVLSTKIEIKLKKPEAVRWEKLEGQGDVPTPKQFVADVKNLYPSSSPYTRNWDKLVG | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 10910 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG3 κ |
| Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA | |
| 6G5 | |
| Western Blot 1:500 | |
| NP_006695 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction