missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUGT1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49707PCP
This item is not returnable.
View return policy
Description
SUGT1 Polyclonal antibody specifically detects SUGT1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SUGT1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| Protein 40-6-3, putative 40-6-3 protein, Sgt1, SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae), SGT1B protein, suppressor of G2 allele of SKP1 homolog, suppressor of G2 allele of SKP1, S. cerevisiae, homolog of | |
| The immunogen for this product maps to a region between residue 315 and 365 of human Suppressor of G2 Allele of SKP1 using the numbering given in entry NP_001124384.1 (GeneID 10910). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10910 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction