missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUGT1 Monoclonal Antibody (OTI10G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810099S
This item is not returnable.
View return policy
Description
SUGT1 Monoclonal antibody specifically detects SUGT1 in Human samples. It is validated for Western BlotSpecifications
| SUGT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y2Z0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10910 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10G2 | |
| Unconjugated | |
| SUGT1 | |
| 2410174K12Rik; 40-6-3; Aa1114; LOC101122608; MGC137150 protein; Protein 40-6-3; Protein SGT1 homolog; putative 40-6-3 protein; Sgt1; SGT1 homolog, MIS12 kinetochore complex assembly cochaperone; SGT1, suppressor of G2 allele of SKP1; SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae); SGT1B protein; SUGT1; suppressor of G2 allele of SKP1; suppressor of G2 allele of SKP1 homolog; suppressor of G2 allele of SKP1, S. cerevisiae, homolog of; suppressor of g2 allele of SKP1-like protein | |
| Full length human recombinant protein of human SUGT1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction