missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUGT1 Monoclonal Antibody (OTI10G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810099
This item is not returnable.
View return policy
Description
This gene is homologous to the yeast gene SGT1, which encodes a protein involved in kinetochore function and required for the G1/S and G2/M transitions. Complementation studies suggest that the human protein has similar functions. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| SUGT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y2Z0 | |
| SUGT1 | |
| Full length human recombit protein of human SUGT1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI10G2 | |
| Unconjugated | |
| SUGT1 | |
| SGT1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10910 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction