missing translation for 'onlineSavingsMsg'
Learn More

SUHW1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-98117JF646

Product Code. 18072318

  • € 415.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SUHW1 Polyclonal specifically detects SUHW1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SUHW1
Polyclonal
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
C19orf37, chromosome 19 open reading frame 37, Enzyme-like protein PIT13, MGC51082, Zfp428, zinc finger protein 428
Produced in rabbits immunized with E. coli-derived Human SUHW1 fragment.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
129025
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.