missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUHW1 Antibody (OTI2H3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74409MFV610
This item is not returnable.
View return policy
Description
SUHW1 Monoclonal antibody specifically detects SUHW1 in Human samples. It is validated for Flow Cytometry
Specifications
| SUHW1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 129025 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI2H3 | |
| Flow Cytometry | |
| C19orf37, chromosome 19 open reading frame 37, Enzyme-like protein PIT13, MGC51082, Zfp428, zinc finger protein 428 | |
| Human recombinant protein fragment corresponding to amino acids 1-333 of human ZNF280A (NP_542778) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction