missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUHW2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05966PCP
This item is not returnable.
View return policy
Description
SUHW2 Polyclonal antibody specifically detects SUHW2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SUHW2 | |
| Immunohistochemistry-Paraffin | |
| Antigen and protein A Affinity-purified | |
| Primary | |
| Purified |
| Polyclonal | |
| Rabbit | |
| RUO | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction