Learn More
Sulcotrione 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30324372
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324372 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30324372 Supplier Dr. Ehrenstorfer Supplier No. DREA16988000AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulcotrione
CAS 99105-77-8
Analyte or Component Names Sulcotrione
Shipping Condition Room Temperature
Molecular Formula C14 H13 Cl O5 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C14H13ClO5S/c1-21(19,20)8-5-6-9(10(15)7-8)14(18)13-11(16)3-2-4-12(13)17/h5-7,13H,2-4H2,1H3
SMILES CS(=O)(=O)c1ccc(C(=O)C2C(=O)CCCC2=O)c(Cl)c1
IUPAC Name 2-(2-chloro-4-methylsulfonylbenzoyl)cyclohexane-1,3-dione
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 328.77
Quantity 1 mL
Formula Weight 328.0172 g/mol