Learn More
Sulcotrione, TRC
Specifications
Specifications
| Chemical Name or Material | Sulcotrione |
| CAS | 99105-77-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H13 Cl O5 S |
| Quantity | 250 mg |
| Synonym | 2-[2-Chloro-4-(methylsulfonyl)benzoyl]-1,3-cyclohexanedione, Chlormesulone, ICI-A 0051, Mikado, SC 0051, Sulcotrione |
| InChI Formula | InChI=1S/C14H13ClO5S/c1-21(19,20)8-5-6-9(10(15)7-8)14(18)13-11(16)3-2-4-12(13)17/h5-7,13H,2-4H2,1H3 |
| SMILES | CS(=O)(=O)c1ccc(C(=O)C2C(=O)CCCC2=O)c(Cl)c1 |
| IUPAC Name | 2-(2-chloro-4-methylsulfonylbenzoyl)cyclohexane-1,3-dione |
| Molecular Weight (g/mol) | 328.77 |
| Show More |