missing translation for 'onlineSavingsMsg'
Learn More

Sulfachloropyridazine, TRC

Product Code. 30417137
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417137 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417137 Supplier Toronto Research Chemicals Supplier No. TRCS68906150MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfachloropyridazine
CAS 80-32-0
Molecular Formula C10 H9 Cl N4 O2 S
Quantity 50 mg
Synonym Benzenesulfonamide','4-amino-N-(6-chloro-3-pyridazinyl)-, Sulfanilamide','N1-(6-chloro-3-pyridazinyl)- (6CI,7CI,8CI), 3-Chloro-6-sulfanilamidopyridazine, 6-Sulfanilamido-3-chloropyridazine, BA 10370, Bactolin, Ciba 10370, Consulid, Cosulid, Durasulf, N1-(6-Chloro-3-pyridazinyl)sulfanilamide, Nefrosul, Renalsulfa, Sonilyn, Sulfachlorpyridazine, Sulfaclorazina, Sulfarene, Sulfochlorpyridazine, Sulpirazina
InChI Formula InChI=1S/C10H9ClN4O2S/c11-9-5-6-10(14-13-9)15-18(16,17)8-3-1-7(12)2-4-8/h1-6H,12H2,(H,14,15)
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2ccc(Cl)nn2
IUPAC Name 4-amino-N-(6-chloropyridazin-3-yl)benzenesulfonamide
Molecular Weight (g/mol) 284.72
Formula Weight 284.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.