missing translation for 'onlineSavingsMsg'
Learn More

Sulfadiazine, 98%

Product Code. 11419513
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11419513

Brand: Thermo Scientific Alfa Aesar A12370.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68-35-9 | C10H10N4O2S | 250.276 g/mol

Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-35-9
Molecular Formula C10H10N4O2S
Molecular Weight (g/mol) 250.276
MDL Number MFCD00006065
InChI Key SEEPANYCNGTZFQ-UHFFFAOYSA-N
Synonym sulfadiazine, sulphadiazine, sulfapyrimidine, sulfadiazin, sulfazine, sulfadiazene, liquadiazine, sulfapyrimidin, adiazin, adiazine
PubChem CID 5215
ChEBI CHEBI:9328
IUPAC Name 4-amino-N-pyrimidin-2-ylbenzenesulfonamide
SMILES C1=CN=C(N=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 253°C to 256°C
Assay Percent Range 99%
Quantity 1000 g
Merck Index 14,8903
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 250.28
Percent Purity 99%
Chemical Name or Material Sulfadiazine
compliance-icons compliance-icons
Product Identifier
  • Sulfadiazine
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.