Learn More
Sulfadimethoxine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfadimethoxine-d4 |
| CAS | 1020719-80-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12H10D4N4O4S |
| Quantity | 10 mg |
| Synonym | Sulfadimethoxine D4 |
| InChI Formula | InChI=1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)/i3D,4D,5D,6D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2cc(OC)nc(OC)n2 |
| Molecular Weight (g/mol) | 314.35 |
| Formula Weight | 314.0987 |
| Show More |