Learn More
Sulfamethazine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfadimidine-d4 |
| CAS | 1020719-82-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 2H4 H10 N4 O2 S |
| Quantity | 10 mg |
| Synonym | Benzene-2,3,5,6-d4-sulfonamide','4-amino-N-(4,6-dimethyl-2-pyrimidinyl)-, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzene-2,3,5,6-d4-sulfonamide, Sulfamethazine-d4, Sulfadimidine-d4, Sulfadimidine (phenyl-D4) |
| InChI Formula | InChI=1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)/i3D,4D,5D,6D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2nc(C)cc(C)n2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 282.35 |
| Show More |