missing translation for 'onlineSavingsMsg'
Learn More

Sulfapyridine, Dr. Ehrenstorfer

Product Code. 30326823
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30326823

Brand: Dr. Ehrenstorfer DREC17000100100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfapyridine
CAS 144-83-2
Analyte or Component Names Sulfapyridine
Shipping Condition +20°C
Molecular Formula C11 H11 N3 O2 S
Certifications/Compliance ISO 17034
Synonym Sulfasalazine Imp. J (EP), Benzenesulfonamide, 4-amino-N-2-pyridinyl-Sulfanilamide, N1-2(1H)-pyridylidene- (6CI,7CI), Sulfanilamide, N1-2-pyridyl- (8CI), Sulfanilamide, N4-2-pyridyl- (4CI), 2-Sulfanilamidopyridine, 2-Sulfapyridine, 4-Amino-N-2-pyridinylbenzenesulfonamide, 4-[(2-Pyridylamino)sulfonyl]aniline, A-499, Adiplon, Coccoclase, Dagenan, Eubasin, Eubasinum, Haptocil, M and B 693, N-(2-Pyridyl)-4-aminobenzenesulfonamide, N1-2-Pyridylsulfanilamide, NSC 41791, NSC 4753, Piridazol, Pirodazole, Plurazol, Pyriamid, Pyridazol, Relbapiridina, Ronin, Septipulmon, Streptosilpyridine, Sulfapyridine, Sulfidin, Sulfidine, Sulphapyridine, Thioseptal, Trianon, Sulfasalazine Impurity J
InChI Formula InChI=1S/C11H11N3O2S/c12-9-4-6-10(7-5-9)17(15,16)14-11-3-1-2-8-13-11/h1-8H,12H2,(H,13,14)
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2ccccn2
IUPAC Name 4-amino-N-(2-pyridyl)benzenesulfonamide
Molecular Weight (g/mol) 249.29
Quantity 100 mg
Formula Weight 249.288 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.