Learn More
Lovibond™ Sulfate Reducing Bacteria (SRB) Tube Test
Product Code. 17880695
Change view
Click to view available options
:
Sulfate Reducing Bacteria (SRB) Tube Test; Detects: Sulfate-reducing Bacteria
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17880695 Sulfate Reducing Bacteria (SRB) Tube Test; Detects: Sulfate-reducing Bacteria
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17880695 Supplier Lovibond™ Supplier No. 56B010810
The Sulfate Reducing Bacteria (SRB) Tube Test is designed to assess the contamination of water samples with sulfate reducing bacteria. The test contains a straw colored medium which reacts to the production of hydrogen sulfide to give a semi-quantitative result after 5 day.
  • The Sulfate Reducing Bacteria (SRB) test is designed to assess the contamination of water samples with sulfate reducing bacteria.
  • The test contains a straw colored medium which reacts to the production of hydrogen sulfide to give a semi-quantitative result after 5 days.
  • The SRB test is used specifically to indicate the presence of bacteria, which under the correct conditions, are able to produce hydrogen sulfide.
  • Hydrogen sulfide is a colorless gas which is extremely corrosive to ferrous and no ferrous metals.
  • This can lead to holes in water systems and leaks in tankers by dissolving the surrounding metal.
Specifications
Detects Sulfate-reducing Bacteria
For Use With (Application) Determine aerobic and anaerobic bacteria levels
Quantity 1 Ea.