Learn More
Sulfiram, Mikromol™
Description
Sulfiram
Specifications
Specifications
| Chemical Name or Material | Sulfiram |
| CAS | 95-05-6 |
| Analyte or Component Names | Sulfiram |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H20 N2 S3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Disulfiram Imp. A (EP), Thiodicarbonic diamide ([(H2N)C(S)]2S), N,N,N',N'-tetraethyl-, Sulfide, bis(diethylthiocarbamoyl) (6CI,7CI,8CI), Thiodicarbonic diamide ([(H2N)C(S)]2S), tetraethyl- (9CI), Thiuram monosulfide, tetraethyl- (4CI), Bis(N,N-diethylthiocarbamoyl) sulfide, Bis(diethylthiocarbamoyl) sulfide, Bis(diethylthiocarbamyl) sulfide, Carbamodithioic acid, diethyl-, anhydrosulfide, Kutka, Kutkasin, Methanethioamide, 1,1'-thiobis[N,N-diethyl-, Monosulfiram, NSC 36731, Sanigal, Sarcocide B, Sulfide, bis[(diethylamino)thioxomethyl], Sulfiram, Sulfirame, Sulfiramum, TTMS, Tetmos, Tetmosol, Tetraethylthiuram monosulfide, Tetrucid, Disulfiram Impurity A |
| InChI Formula | InChI=1S/C10H20N2S3/c1-5-11(6-2)9(13)15-10(14)12(7-3)8-4/h5-8H2,1-4H3 |
| SMILES | CCN(CC)C(=S)SC(=S)N(CC)CC |
| IUPAC Name | diethylcarbamothioyl N,N-diethylcarbamodithioate |
| Show More |