Learn More
Sulfiram, TRC
Product Code. 30004872
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004872 5 g 5g
17993045 250 mg 250mg
17901655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004872 Supplier Toronto Research Chemicals Supplier No. TRCS7335005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-05-6
Molecular Formula C10 H20 N2 S3
Molecular Weight (g/mol) 264.47
Synonym Thiodicarbonic diamide ([(H2N)C(S)]2S), N,N,N',N'-tetraethyl-, Sulfide, bis(diethylthiocarbamoyl) (6CI,7CI,8CI), Thiodicarbonic diamide ([(H2N)C(S)]2S), tetraethyl- (9CI), Thiuram monosulfide, tetraethyl- (4CI), Bis(N,N-diethylthiocarbamoyl) sulfide, Bis(diethylthiocarbamoyl) sulfide, Bis(diethylthiocarbamyl) sulfide, Carbamodithioic acid, diethyl-, anhydrosulfide, Kutka, Kutkasin, Methanethioamide, 1,1'-thiobis[N,N-diethyl-, Monosulfiram, NSC 36731, Sanigal, Sarcocide B, Sulfide, bis[(diethylamino)thioxomethyl], Sulfiram, Sulfirame, Sulfiramum, TTMS, Tetmos, Tetmosol, Tetraethylthiuram monosulfide, Tetrucid
IUPAC Name diethylcarbamothioyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SC(=S)N(CC)CC
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H20N2S3/c1-5-11(6-2)9(13)15-10(14)12(7-3)8-4/h5-8H2,1-4H3
Formula Weight 264.0789
Percent Purity >95%
Chemical Name or Material Sulfiram
Grade HPLC