Learn More
Invitrogen™ Sulfotyrosine Monoclonal Antibody (7C5), PerCP
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545835
Product Code. 17848452
  • € 481.00 / 100µg
Description
A 1:1,000 dilution of MA5-45835 was sufficient for detection of Sulfotyrosine in 0.5 μg of Sulfotyrosine conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine.
Sulfotyrosine is involved in protein-protein interactions. Sulfotyrosines are present in all multicellular eukaryotes, but unicellular eukaryotes such as yeast and prokaryotes are not capable of tyrosine sulfation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sulfotyrosine
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Mouse
Protein G
RUO
Chemical
Antibody
IgG1
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
7C5
PerCP
sTYR; S-Tyrosine; Sulfated Tyrosine; Sulfotyrosine; Sulphated Tyrosine; Sulphotyrosine; Tyrosine sulfation; Tyrosine Sulphation
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μg
Primary
4°C
Liquid
Chemical