Learn More
Sulfur in Diesel Fuel standard solution, Specpure™, 20μg/g (0.0020%)
Product Code. 15486885
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 15486885
Brand: Thermo Scientific Alfa Aesar 045311.AE
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Density 0.6 to 0.9 g/mL
Flash Point 69°C (156°F)
Physical Form Liquid
MDL Number MFCD01095233
Color Brown
Boiling Point 171°C to 315°C
Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Diesel Fuel standard solution, 20μg/g (0.0020%)
compliance-icons
Product Identifier
  • Sulfur in Diesel Fuel standard solution, Specpure, 20 µg/g (0.0020%)
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Fuels, diesel, no. 2