Learn More
Sulindac, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Sulindac |
| CAS | 38194-50-2 |
| Analyte or Component Names | Sulindac |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H17 F O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sulindac, (1Z)-5-Fluoro-2-methyl-1-[[4-(methylsulfinyl)phenyl]methylene]-1H-indene-3-acetic acid, cis-5-fluoro-2-methyl-1-[p-(methylsulfinyl)benzylidene]indene-3-acetic acid, MK-231, Aflodac, Algocetil, Arthrocine, Artribid, Citireuma, Clinoril, Clisundac, Reumofil, Reumyl, Sudac, Sulinol, Sulreuma |
| InChI Formula | InChI=1S/C20H17FO3S/c1-12-17(9-13-3-6-15(7-4-13)25(2)24)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9- |
| Solution Type | Neat |
| SMILES | CC1=C(CC(=O)O)c2cc(F)ccc2/C/1=C\c3ccc(cc3)S(=O)C |
| Show More |