Learn More
Sulindac, TRC
Product Code. 30136127
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136127 1 g 1g
30132845 5 g 5g
17969849 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136127 Supplier Toronto Research Chemicals Supplier No. TRCS6992151G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38194-50-2
Molecular Formula C20 H17 F O3 S
Molecular Weight (g/mol) 356.41
Synonym Sulindac, (1Z)-5-Fluoro-2-methyl-1-[[4-(methylsulfinyl)phenyl]methylene]-1H-indene-3-acetic acid, cis-5-fluoro-2-methyl-1-[p-(methylsulfinyl)benzylidene]indene-3-acetic acid, MK-231, Aflodac, Algocetil, Arthrocine, Artribid, Citireuma, Clinoril, Clisundac, Reumofil, Reumyl, Sudac, Sulinol, Sulreuma
IUPAC Name 2-[(3Z)-6-fluoro-2-methyl-3-[(4-methylsulfinylphenyl)methylidene]inden-1-yl]acetic acid
SMILES CC1=C(CC(=O)O)c2cc(F)ccc2/C/1=C\c3ccc(cc3)S(=O)C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C20H17FO3S/c1-12-17(9-13-3-6-15(7-4-13)25(2)24)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9-
Formula Weight 356.0882
Chemical Name or Material Sulindac