missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Sulphate Cuvette Tests

Product Code. 10192014
Change view
Click to view available options
Range:
150 to 900mg/L SO4
40 to 150mg/L SO4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10192014 40 to 150mg/L SO4 Each
10686213 150 to 900mg/L SO4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192014 Supplier Hach Lange™ Supplier No. LCK153

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

No. of Tests 25
Range 40 to 150mg/L SO4
Storage Requirements 15°C to 25°C
Test Method Barium Sulphate
Type Sulphate Cuvette Test
compliance-icons
Product Identifier
  • Sulfat/Sulphate/Sulfate, LCK 153 A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.