Learn More
Hach Lange™ Sulphate Cuvette Tests
Product Code. 10686213
Change view
Click to view available options
Range:
150 to 900mg/L SO4
40 to 150mg/L SO4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10686213 150 to 900mg/L SO4 Each
10192014 40 to 150mg/L SO4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10686213 Supplier Hach Lange™ Supplier No. LCK353
Specifications
No. of Tests 25
Range 150 to 900mg/L SO4
Storage Requirements 15°C to 25°C
Test Method Barium Sulphate
Type Sulphate Cuvette Test
compliance-icons compliance-icons
Product Identifier
  • Sulfat/Sulphate/Sulfate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Glycerol, n-propanol, Hydrochloric acid