missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulpiride EP Impurity G, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Sulpiride EP Impurity G
Specifications
Specifications
| Chemical Name or Material | O-Desmethylsulpiride |
| CAS | 67381-52-6 |
| Analyte or Component Names | O-Desmethylsulpiride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H21 N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | (RS)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulphamoylbenzamide, Sulpiride Imp. G (EP), O-Desmethylsulpiride, Sulpiride Impurity G |
| InChI Formula | InChI=1S/C14H21N3O4S/c1-2-17-7-3-4-10(17)9-16-14(19)12-8-11(22(15,20)21)5-6-13(12)18/h5-6,8,10,18H,2-4,7,9H2,1H3,(H,16,19)(H2,15,20,21) |
| SMILES | CCN1CCCC1CNC(=O)c2cc(ccc2O)S(=O)(=O)N |
| IUPAC Name | N-[(1-ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulfamoylbenzamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction