missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1A1 Monoclonal Antibody (OTI8B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501820
This item is not returnable.
View return policy
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene.Specifications
| SULT1A1 | |
| Monoclonal | |
| 0.71 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50225 | |
| SULT1A1 | |
| Full length human recombit protein of human SULT1A1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI8B8 | |
| Unconjugated | |
| SULT1A1 | |
| HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6817 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction